2-Amino-5-methoxypyridine ≥95%

Supplier: Matrix Scientific

073245-1G 073245-5G
102529-152EA 90.77 USD
102529-152 102529-154
2-Amino-5-methoxypyridine ≥95%
2-Amino-5-methoxypyridine

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.1405 g/mol
MDL Number: MFCD07374873
CAS Number: 10167-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR