1H-Pyrazole-1-carboximidamide hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

073215-100G
102528-992EA 200 USD
102528-992
1H-Pyrazole-1-carboximidamide hydrochloride ≥95%
1H-Pyrazole-1-carboximidamide hydrochloride

Hazard: Irritant

Formula: C₄H₇ClN₄
MW: 146.58 g/mol
Melting Pt: 165…166 °C
MDL Number: MFCD00210087
CAS Number: 4023-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR