4-Hydroxy-2-(methylthio)pyrimidine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

073212-1G 073212-5G 073212-10G
102528-970EA 130.77 USD
102528-970 102528-972 102528-974
4-Hydroxy-2-(methylthio)pyrimidine-5-carboxylic acid ≥95%
4-Hydroxy-2-(methylthio)pyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₆N₂O₃S
MDL Number: MFCD09759031
CAS Number: 397308-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR