1,4-Dioxaspiro[4.5]decan-8-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

073152-100G
102528-658EA 158.46 USD
102528-658
1,4-Dioxaspiro[4.5]decan-8-one ≥95%
1,4-Dioxaspiro[4.5]decan-8-one

Hazard: Irritant/Moisture Sensitive

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 72…75 °C
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR