6-Bromo-1-indanone ≥95%

Supplier: Matrix Scientific

073015-5G
102527-934EA 29.23 USD
102527-934
6-Bromo-1-indanone ≥95%
6-Bromo-1-indanone

Hazard: Irritant

Formula: C₉H₇BrO
MDL Number: MFCD02179286
CAS Number: 14548-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR