3-Acetyl-5-bromopyridine ≥95%

Supplier: Matrix Scientific

072837-5G
102527-012EA 138.46 USD
102527-012
3-Acetyl-5-bromopyridine ≥95%
3-Acetyl-5-bromopyridine

Hazard: Irritant

Formula: C₇H₆BrNO
MDL Number: MFCD03086033
CAS Number: 38940-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR