2-Pyrrolecarbaldehyde ≥95%

Supplier: Matrix Scientific

072683-10G
102526-168EA 44.62 USD
102526-168
2-Pyrrolecarbaldehyde ≥95%
2-Pyrrolecarbaldehyde

Hazard: Irritant

Formula: C₅H₅NO
Melting Pt: 40…46 °C
MDL Number: MFCD00005217
CAS Number: 1003-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR