2-Deoxy-D(-)-ribose ≥95%

Supplier: Matrix Scientific

072649-100G
102525-996EA 260 USD
102525-996
2-Deoxy-D(-)-ribose ≥95%
2-Deoxy-D(-)-ribose

TCA listed: T
Hazard: Irritant

Formula: C₅H₁₀O₄
Melting Pt: 76…85 °C
MDL Number: MFCD00135904
CAS Number: 533-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR