2-Amino-5-bromo-3-methylpyrazine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-3-methylpyrazin-2-amine
072590-250MG
072590-1G
102525-678EA
826.15
USD
102525-678
102525-680
2-Amino-5-bromo-3-methylpyrazine ≥95%
2-Amino-5-bromo-3-methylpyrazine
Hazard: Irritant
Formula:
C₅H₆BrN₃ |
MDL Number:
MFCD08705773 CAS Number: 74290-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...