4-Aminopyrimidine ≥95%

Supplier: Matrix Scientific

072535-5G
102525-370EA 113.85 USD
102525-370
4-Aminopyrimidine ≥95%
4-Aminopyrimidine

Hazard: Irritant

Formula: C₄H₅N₃
MW: 95.1
Melting Pt: 147…152 °C
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR