3-Benzyl-7-chloro-3H-[1,2,3]triazolo[4,5-d]pyrimidine ≥95%

Supplier: Matrix Scientific

072379-250MG 072379-1G 072379-5G
102524-534EA 640 USD
102524-534 102524-536 102524-538
3-Benzyl-7-chloro-3H-[1,2,3]triazolo[4,5-d]pyrimidine ≥95%
3-Benzyl-7-chloro-3H-[1,2,3]triazolo[4,5-d]pyrimidine

Hazard: Irritant

Formula: C11H8ClN5
MW: 245.6700 g/mol
MDL Number: MFCD09038669
CAS Number: 21410-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR