2-Formyl-4-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methylpyridine-2-carboxaldehyde, 4-Methylpicolinaldehyde

072334-1G 072334-5G
102524-306EA 150.77 USD
102524-306 102524-308
2-Formyl-4-picoline ≥95%
2-Formyl-4-picoline

Hazard: Irritant

Formula: C₇H₇NO
Boiling Pt: 94…97 °C (15 Torr)
MDL Number: MFCD07367933
CAS Number: 53547-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR