2-Methylpyrimidine-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

072118-250MG 072118-1G 072118-5G
102523-188EA 569.23 USD
102523-188 102523-190 102523-192
2-Methylpyrimidine-4-carbaldehyde ≥95%
2-Methylpyrimidine-4-carbaldehyde

Hazard: Irritant

Formula: C₆H₆N₂O
MDL Number: MFCD09832936
CAS Number: 1004-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR