2-chloro-5-iodopyrimidine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-iodopyrimidine
072113-1G
072113-5G
102523-162EA
89.23
USD
102523-162
102523-164
2-chloro-5-iodopyrimidine ≥95%
2-chloro-5-iodopyrimidine
Hazard: Irritant
Formula:
C₄H₂ClIN₂ |
MDL Number:
MFCD06200210 CAS Number: 32779-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...