Ethyl-2-(4-fluorophenyl)thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

072056-1G 072056-5G
102522-868EA 710.77 USD
102522-868 102522-870
Ethyl-2-(4-fluorophenyl)thiazole-4-carboxylate ≥95%
Ethyl-2-(4-fluorophenyl)thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀FNO₂S
MDL Number: MFCD06738337
CAS Number: 132089-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR