5-Nitro-1H-indazole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

072046-1G 072046-5G 072046-10G
102522-818EA 335.38 USD
102522-818 102522-820 102522-822
5-Nitro-1H-indazole-3-carbaldehyde ≥95%
5-Nitro-1H-indazole-3-carbaldehyde

Hazard: Irritant

Formula: C₈H₅N₃O₃
MDL Number: MFCD07781436
CAS Number: 677702-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR