7-Methyl-1H-indazole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

072045-1G 072045-5G 072045-10G
102522-812EA 427.69 USD
102522-812 102522-814 102522-816
7-Methyl-1H-indazole-3-carbaldehyde ≥95%
7-Methyl-1H-indazole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₈N₂O
MW: 160.18 g/mol
MDL Number: MFCD09027036
CAS Number: 1000340-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR