5-Bromo-2-(3-methoxypropylamino)pyrimidine ≥95%

Supplier: Matrix Scientific

072043-1G 072043-5G 072043-10G
102522-800EA 355.38 USD
102522-800 102522-802 102522-804
5-Bromo-2-(3-methoxypropylamino)pyrimidine ≥95%
5-Bromo-2-(3-methoxypropylamino)pyrimidine

Hazard: Irritant

Formula: C₈H₁₂BrN₃O
MW: 246.11 g/mol
MDL Number: MFCD09743709
CAS Number: 1189482-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR