4-(Thiazol-2-yloxy)phenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Thiazol-2-yloxy)aniline

072037-1G 072037-5G 072037-10G
102522-764EA 276.92 USD
102522-764 102522-766 102522-768
4-(Thiazol-2-yloxy)phenylamine ≥95%
4-(Thiazol-2-yloxy)phenylamine

Hazard: Irritant

Formula: C₉H₈N₂OS
MW: 192.24 g/mol
MDL Number: MFCD09743704
CAS Number: 105350-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR