2,3,4,5-Tetrahydro-1H-benzo[b]azepine ≥95%

Supplier: Matrix Scientific

071962-1G 071962-5G 071962-10G
102522-336EA 355.38 USD
102522-336 102522-338 102522-340
2,3,4,5-Tetrahydro-1H-benzo[b]azepine ≥95%
2,3,4,5-Tetrahydro-1H-benzo[b]azepine

Hazard: Irritant

Formula: C₁₀H₁₃N
MDL Number: MFCD00272363
CAS Number: 1701-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR