2,3,4,5-Tetrahydro-1H-benzo[b]azepine ≥95%
Supplier: Matrix Scientific
071962-1G
071962-5G
071962-10G
102522-336EA
355.38
USD
102522-336
102522-338
102522-340
2,3,4,5-Tetrahydro-1H-benzo[b]azepine ≥95%
2,3,4,5-Tetrahydro-1H-benzo[b]azepine
Hazard: Irritant
Formula:
C₁₀H₁₃N |
MDL Number:
MFCD00272363 CAS Number: 1701-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...