2-(3-Methoxyphenyl)pyrimidine-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

071939-1G 071939-5G
102522-206EA 710.77 USD
102522-206 102522-208
2-(3-Methoxyphenyl)pyrimidine-5-carbaldehyde ≥95%
2-(3-Methoxyphenyl)pyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
CAS Number: 1119398-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR