2'-(Trifluoromethyl)biphenyl-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

071877-1G 071877-5G 071877-10G
102521-854EA 355.38 USD
102521-854 102521-856 102521-858
2'-(Trifluoromethyl)biphenyl-3-carboxylic acid ≥95%
2'-(Trifluoromethyl)biphenyl-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉F₃O₂
MDL Number: MFCD03424602
CAS Number: 168618-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR