4-(2-Pyridyl)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde

071847-1G 071847-5G 071847-10G
102521-682EA 72.31 USD
102521-682 102521-684 102521-686
4-(2-Pyridyl)benzaldehyde ≥95%
4-(2-Pyridyl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₉NO
MDL Number: MFCD01863537
CAS Number: 127406-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR