[2-(4-Ethylpiperazin-1-yl)phenyl]methanol ≥95%
Supplier: Matrix Scientific
071845-1G
071845-5G
071845-10G
102521-670EA
284.62
USD
102521-670
102521-672
102521-674
[2-(4-Ethylpiperazin-1-yl)phenyl]methanol ≥95%
[2-(4-Ethylpiperazin-1-yl)phenyl]methanol
Hazard: Irritant
Formula:
C₁₃H₂₀N₂O |
CAS Number:
914349-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...