4-Dimethoxymethylpyrimidin-2-ylamine ≥95%

Supplier: Matrix Scientific

071794-1G 071794-5G 071794-10G
102521-374EA 269.23 USD
102521-374 102521-376 102521-378
4-Dimethoxymethylpyrimidin-2-ylamine ≥95%
4-Dimethoxymethylpyrimidin-2-ylamine

Hazard: Irritant

Formula: C₇H₁₁N₃O₂
MDL Number: MFCD05864792
CAS Number: 165807-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR