4-(4-Ethylpiperazin-1-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

071679-1G 071679-5G 071679-10G
102520-708EA 249.23 USD
102520-708 102520-710 102520-712
4-(4-Ethylpiperazin-1-yl)benzaldehyde ≥95%
4-(4-Ethylpiperazin-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MDL Number: MFCD05864670
CAS Number: 197638-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR