(4S)-3-[(2E)-3-(4-Methylphenyl)prop-2-enoyl]-4-phenyl-1,3-oxazolidin-2-one ≥97%

Supplier: Matrix Scientific

071367-500MG 071367-1G
102519-914EA 403.08 USD
102519-914 102519-916
(4S)-3-[(2E)-3-(4-Methylphenyl)prop-2-enoyl]-4-phenyl-1,3-oxazolidin-2-one ≥97%
(4S)-3-[(2E)-3-(4-Methylphenyl)prop-2-enoyl]-4-phenyl-1,3-oxazolidin-2-one

Hazard: Irritant

Formula: C₁₉H₁₇NO₃
MDL Number: MFCD22126134
CAS Number: 915390-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR