3-(3-Fluoro-4-methylphenyl)-1,2,4-oxadiazole ≥97%

Supplier: Matrix Scientific

071345-500MG 071345-1G
102519-824EA 298.46 USD
102519-824 102519-826
3-(3-Fluoro-4-methylphenyl)-1,2,4-oxadiazole ≥97%
3-(3-Fluoro-4-methylphenyl)-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₉H₇FN₂O
MDL Number: MFCD13181200
CAS Number: 1146699-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR