1-Chloro-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole ≥97%

Supplier: Matrix Scientific

071221-5G
102519-630EA 1592.31 USD
102519-630
1-Chloro-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole ≥97%
1-Chloro-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole

Hazard: Irritant

Formula: C₉H₁₀ClIO
Melting Pt: 148…150 °C (dec.)
MDL Number: MFCD21648251
CAS Number: 69352-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR