3-Aminobutan-1-ol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-butan-1-ol
061407-1G
061407-5G
102518-888EA
15.38
USD
102518-888
102833-312
3-Aminobutan-1-ol ≥95%
3-Aminobutan-1-ol
Hazard: Irritant
Formula:
C₄H₁₁NO |
MDL Number:
MFCD03412694 CAS Number: 2867-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...