Methyl-4-bromo-3-nitrobenzoate

Supplier: Matrix Scientific

Synonyms: 4-Bromobenzoic acid methyl ester, Methyl 4-bromo-3-nitrobenzoate

057604-5G 057604-25G
102518-852EA 93.85 USD
102518-852 102518-854
Methyl-4-bromo-3-nitrobenzoate
Methyl 4-bromo-3-nitrobenzoate

Hazard: Irritant

Formula: C₈H₆BrNO₄
MDL Number: MFCD00553089
CAS Number: 2363-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR