JC-1 iodide salt ≥98%

Supplier: 0000027161

Synonyms: 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, JC-1, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, (E)-5,6-Dichloro-2-(3-(5,6-dichloro-1,3-diethyl-1,3-dihydro-2H-benzo[d]imidazol-2-ylidene)prop-1-en-1-yl)-1,3-diethyl-1H-benzo[d]imidazol-3-ium iodide, CBIC2(3), 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide

AG-CR1-3568-M001 AG-CR1-3568-M005 AG-CR1-3568-5001
102516-308EA 76.15 USD
102516-308 102516-310 102516-312
JC-1 iodide salt ≥98%
JC-1 iodide salt

The membrane-permeant dual-emission potential-sensitive JC-1 dye is widely used in apoptosis studies to monitor mitochondrial health by flow cytometry, fluorescence microscopy and in microplate-based fluorescent assays. JC-1 dye can be used as an indicator of mitochondrial membrane potential in a variety of cell types, including myocytes and neurons, as well as in intact tissues and isolated mitochondria. JC-1 accumulates in mitochondria, selectively generating an orange J-aggregate emission profile (590 nm) in healthy cells. After cell injury, as membrane potential decreases, JC-1 monomers are generated, resulting in a shift to green emission (529 nm). The principal advantage of JC-1 relative to other commonly employed fluorescent probes of mitochondrial membrane potential is that it allows qualitative visualization, considering the shift from orange to green fluorescence emission, and quantitative detection, considering the fluorescence intensity ratio.

Formula: C₂₅H₂₇Cl₄IN₄
Storage Temperature: Freezer
MDL Number: MFCD00188475
CAS Number: 47729-63-5

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Red powder.
Solubility Soluble in DMSO, ethanol, methanol or acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR