3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98%

Supplier: 0000027161

Synonyms: 3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3,4-Bis(3-indolyl)maleimide, Bisindolylmaleimide IV, Arcyriarubin A

AG-CR1-0152-M001 AG-CR1-0152-M005
102516-208EA 76.15 USD
102516-208 102516-210
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98%
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione

Cell permeable protein kinase C (PKC) inhibitor. Inhibits also protein kinase A (PKA).

Formula: C20H13N3O2
Storage Temperature: Freezer
MDL Number: MFCD00236432
CAS Number: 119139-23-0

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Red to orange solid.
Solubility Soluble in DMSO, ethanol, methanol or dimethylformamide.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR