YC-1 ≥98%

Supplier: 0000027161

Synonyms: (5-(1-Benzyl-1H-indazol-3-yl)furan-2-yl)methanol, Lificiguat

AG-CR1-0120-M001 AG-CR1-0120-M005 AG-CR1-0120-M025
102516-188EA 27.69 USD
102516-188 102516-190 102516-192
YC-1 ≥98%
YC-1

Nitric oxide (NO) independent, superoxide-sensitive soluble guanylyl cyclase (sGC) activator. Induces concentration-dependent increase in cGMP levels. Inhibits platelet adhesion to collagen. Thombosis inhibitor. Non-specific phosphodiesterase inhibitor. Na+ channel blocker. Anticancer compound. HIF-alpha inhibitor. Blocks angiogenesis. Tumor growth inhibitor. Apoptosis inducer. NK cell differentiation enhancer. TRAIL sensitizer.

Formula: C₁₉H₁₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD06407798
CAS Number: 170632-47-0

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to pale yellow powder.
Solubility Soluble in DMSO, methanol or dichloromethane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR