Bisindoylmaleimide X hydrochloride ≥98%
Supplier: 0000027161
Synonyms:
Bisindolylmaleimide X HCl, Ro 31-8425, 3-(8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione hydrochloride
AG-CR1-0113-M001
AG-CR1-0113-M005
102516-166EA
96.92
USD
102516-166
102516-168
Bisindoylmaleimide X hydrochloride ≥98%
Bisindoylmaleimide X hydrochloride
Selective protein kinase C (PKC) inhibitor. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2). Inhibits superoxide generation in human neutrophils.
Formula:
C₂₆H₂₄N₄O₂·HCl Storage Temperature: Freezer |
MDL Number:
MFCD04972010 CAS Number: 145317-11-9 |
Specification Test Results
Purity |
|
Appearance/Color | Red solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...