2-Morpholino-8-phenylchromone ≥99%

Supplier: 0000027161

Synonyms: LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran

AG-CR1-0108-M001 AG-CR1-0108-M005 AG-CR1-0108-M025
102516-144EA 34.62 USD
102516-144 102516-146 102516-148
2-Morpholino-8-phenylchromone ≥99%
2-Morpholino-8-phenylchromone

Potent, cell permeable, highly specific, reversible PI(3)K (phosphoinositide 3-kinase) inhibitor. pan-class I / II / III PI3K inhibitor. Acts on the ATP-binding site of the enzyme. In solution more stable than wortmannin. Antagonizes P-glycoprotein-mediated multidrug resistance. Blocks Akt phosphorylation. Pim-1 kinase inhibitor. Autophagy inhibitor (autophagosome formation). Inhibitor of BET bromodomain proteins BRD2, BRD3 and BRD4.

Formula: C₁₉H₁₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD00270881
CAS Number: 154447-36-6

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color White to off-white crystalline solid.
Solubility Soluble in DMSO, 100% ethanol or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR