bpV(HOpic) ≥95%
Supplier: 0000027161
AG-CR1-0044-M005
AG-CR1-0044-M025
102516-052EA
96.92
USD
102516-052
102516-054
bpV(HOpic) ≥95%
bpV(HOpic)
Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic. PTEN inhibitor.
Formula:
K₂[VO(O₂)₂C₆H₄NO₃] Storage Temperature: Freezer |
CAS Number:
722494-26-0 |
Specification Test Results
Purity |
|
Appearance/Color | Yellow crystalline solid. |
Solubility | Soluble in DMSO or water. NOTE: The product is not stable in water. Decomposition starts immediately after solubilization. Rate of decomposition is slower in dry DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...