bpV(HOpic) ≥95%

Supplier: 0000027161

AG-CR1-0044-M005 AG-CR1-0044-M025
102516-052EA 96.92 USD
102516-052 102516-054
bpV(HOpic) ≥95%
bpV(HOpic)

Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic. PTEN inhibitor.

Formula: K₂[VO(O₂)₂C₆H₄NO₃]
Storage Temperature: Freezer
CAS Number: 722494-26-0

Order Now

Specification Test Results

Purity 95% (51V-NMR)
Appearance/Color Yellow crystalline solid.
Solubility Soluble in DMSO or water. NOTE: The product is not stable in water. Decomposition starts immediately after solubilization. Rate of decomposition is slower in dry DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR