SB202190 ≥97% (by HPLC)

Supplier: 0000027161
Warning

AG-CR1-0028-M001 AG-CR1-0028-M005 AG-CR1-0028-M025
102515-978EA 41.54 USD
102515-978 102515-980 102515-982
SB202190 ≥97% (by HPLC)
SB202190

Potent and cell permeable p38 MAP kinase inhibitor. Apoptosis inducer. Inhibits p38alpha and beta, but not gamma and delta isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. JNK activator. Autophagic vacuole inducer

Formula: C₂₀H₁₄FN₃O
Storage Temperature: Freezer
MDL Number: MFCD00941964
CAS Number: 152121-30-7

Order Now

Specification Test Results

Purity 97% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in methanol, DMSO or acetone; slightly soluble in ethyl acetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR