MK-571 sodium salt ≥97% (by NMR)

Supplier: 0000027161
Warning

AG-CR1-0021-M005 AG-CR1-0021-M025
102515-946EA 69.23 USD
102515-946 102515-948
MK-571 sodium salt ≥97% (by NMR)
MK-571 sodium salt

Selective leukotriene LTD4 receptor antagonist. Potent MRP1 inhibitor. Antinociceptive.

Formula: C₂₆H₂₆ClN₂O₃S₂·Na
Storage Temperature: Freezer
MDL Number: MFCD11114392
CAS Number: 115103-85-0

Order Now

Specification Test Results

Purity 97% (NMR)
Appearance/Color Light yellow solid.
Solubility Soluble in water (20 mg/ml), 100% ethanol (10 mg/ml), DMSO or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR