Nocodazole ≥98% (by NMR)

Supplier: 0000027161
Warning

AG-CR1-0019-M005 AG-CR1-0019-M010 AG-CR1-0019-M025 AG-CR1-0019-M050
102515-934EA 48.46 USD
102515-934 102515-936 102515-938 102515-940
Nocodazole ≥98% (by NMR)
Nocodazole

Microtubule inhibitor. Antitumor compound. Mitosis inhibitor. Arrests the cell cycle at G2/M phase. Promotes tubulin depolymerization. Induces fragmentation of the Golgi complex. Inhibits the T cell antigen receptor. Stimulates the intrinsic GTPase activity of tubulin. Activates the JNK/SAPK signaling pathway. Apoptosis inducer. Autophagy inhibition through prevention of autophagosome-lysosome fusion.

Formula: C₁₄H₁₁N₃O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00005588
CAS Number: 31430-18-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or dimethylformamide; almost insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR