Avarol ≥97%

Supplier: 0000027161

AG-CN2-0044-M001
102515-828EA 221.54 USD
102515-828
Avarol ≥97%
Avarol

Antibacterial against selected Gram-positive strains, antifungal. Anticancer compound. Antileukemic. Antimitotic agent. Antiviral compound. Inhibits HIV activity. Anti-inflammatory. Synovial phospholipase A2, cyclooxygenase and lipoxygenase inhibitor. Modulator of superoxide dismutase and glutathione peroxidase. Platelet aggregation inhibitor. Anti-psoriatic, mediated by inhibition of TNF-alpha generation and NF-kappaB activation. Reviews.

Formula: C₂₁H₃₀O₂
Storage Temperature: Freezer
CAS Number: 55303-98-5

Order Now

Specification Test Results

Source/Host Isolated from sponge Dysidea avara.
Purity 97% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in ethanol or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR