Ingenol-5,20-acetonide ≥95% (by HPLC)

Supplier: 0000027161

AG-CN2-0029-M001 AG-CN2-0029-M005
102515-776EA 131.54 USD
102515-776 102515-778
Ingenol-5,20-acetonide ≥95% (by HPLC)
Ingenol-5,20-acetonide

Intermediate and starting material for the synthesis and preparation of ingenol 3-ester derivatives. For use in chemical synthesis see references. Has improved shelf life compared to ingenol.

Formula: C₂₃H₃₂O₅
Storage Temperature: Freezer
CAS Number: 77573-43-4

Order Now

Specification Test Results

Source/Host Semisynthetic.
Purity 95% (HPLC)
Appearance/Color White solid.
Solubility Soluble in methanol, DMSO or dichloromethane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR