Staurosporine (from Streptomyces species) ≥98% (by HPLC)

Supplier: 0000027161

Synonyms: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282

AG-CN2-0022-C100 AG-CN2-0022-C500 AG-CN2-0022-M001 AG-CN2-0022-M005
102515-738EA 34.62 USD
102515-738 102515-740 102515-742 102515-744
Staurosporine (from Streptomyces species) ≥98% (by HPLC)
Staurosporine

Staurosporine inhibits platelet aggregation induced by collagen or ADP. It is isolated from Streptomyces spiralis.


  • Antifungal and anti-yeast activity
  • Potent, cell permeable, reversible, ATP-competitive and broad spectrum inhibitor of protein kinases
  • Key apoptosis inducer


Antibiotic. Has no effect on thrombin-induced platelet aggregation. Inhibits protein kinase A, CaM kinase, myosin light chain kinase, protein kinase C, protein kinase G, CDK1/cyclin B, CDK2/cyclin A, CDK4/cyclin D, CDK5/p25, GSK-3beta and Pim-1 kinase. Key apoptosis inducer. Topoisomerase II (Topo II) inhibitor.


Off-white to yellow solid


Stable for at least 2 years after receipt when stored at +4°C.


Soluble in DMSO, ethyl acetate or dimethyl formamide. Insoluble in water.

Formula: C₂₈H₂₆N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD18252446
CAS Number: 62996-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR