Radicicol (from Gilmaniella species) ≥99% (by HPLC)
Supplier: ADIPOGEN CORP MS
Radicicol is an antibiotic, protein tyrosine kinase inhibitor.
- Cyclooxygenase-2 (COX-2) expression inhibitor
- Suppresses expression of iNOS (NOS II)
- Apoptosis inducer
Antibiotic. Protein tyrosine kinase inhibitor. Has anti-fungal, anti-malarial, anti-angiogenic (in vivo), anti-inflammatory and anti-viral activity. Cyclooxygenase-2 (COX-2) expression inhibitor without affecting COX-1 expression in LPS-stimulated macrophages. Induces the differentiation of HL-60 cells into macrophages. Potent HSP90 inhibitor. Anticancer compound. In vivo Rad/Raf interaction inhibitor. Inhibitor of AP-1-, NF-kappaB- and serum response factor (SRF)-mediated transcription. Non-competitive inhibitor of ATP citrate lyase. DNA topoisomerase VI and type II DNA topoisomerase inhibitor. Protects against LPS/IFN-gamma-induced neuronal cell death.
White to slightly yellow solid
Stable for at least two years after receipt when stored at -20°C. Store solutions at -20°C in the dark.
Soluble in DMSO, methanol, acetone, ethyl acetate or chloroform.
Formula:
C₁₈H₁₇ClO₆ Storage Temperature: Freezer |
CAS Number:
12772-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...