K-252a (from Nonomuraea longicatena chiba) ≥98% (by HPLC)

Supplier: 0000027161

Synonyms: Methyl-6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo[b,h]cycloocta[1,2,3,4-jkl]cyclopenta[e]-as-indacene-6-carboxylate

AG-CN2-0019-C100 AG-CN2-0019-M001
102515-726EA 83.08 USD
102515-726 102515-728
K-252a (from Nonomuraea longicatena chiba) ≥98% (by HPLC)
K-252a

K-252a is an antibiotic, PKC, PKA and PKG inhibitor.


  • Highly potent cell permeable CaM kinase and phosphorylase inhibitor
  • Apoptosis inducer
  • Potential drug for psoriasis treatment


Antibiotic. PKC, PKA and PKG inhibitor. Shows neurotropic activity by inducing neurite outgrowth. Tyrosine protein kinase inhibitor of the TRK family. Antiproliferative. Cell cycle arrest inducer.


White to off-white powder


Soluble in DMSO, methanol, methylene chloride, ethyl acetate or acetone. Insoluble in water.

Formula: C₂₇H₂₁N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00161522
CAS Number: 97161-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR