Ferutinin ≥98%, high purity

Supplier: 0000027161

AG-CN2-0007-M001 AG-CN2-0007-M005 AG-CN2-0007-M010
102515-678EA 55.38 USD
102515-678 102515-680 102515-682
Ferutinin ≥98%, high purity
Ferutinin

Potent, naturally occuring non-steroid estrogenic compound. Tool to promote mitochondrial calcium overload and to promote calcium-dependent opening of the mitochondrial permeability transition pore (mPTP). Strong agonist for estrogen receptor (ER)alpha and agonist/antagonist for ERbeta. Calcium ionophoretic Apoptosis modulator. Antiproliferative. Increases nitric oxide synthase activity and phosphoinositides breakdown in nervous tissue. Shows aphrodisiac and anti-sexual impotence activities. Anti-osteoporotic. Prevents osteoporosis caused by severe estrogen deficiency. Modest AChE inhibitor.

Formula: C₂₂H₃₀O₄
Storage Temperature: Freezer
CAS Number: 41743-44-6

Order Now

Specification Test Results

Source/Host Semisynthetic.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, acetone, dichloromethane or ethyl acetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR