Human Recombinant BCMA (from HEK293 cells)
Supplier: 0000027161
AG-40B-0080-C050 AG-40B-0080-3050
102515-556EA 332.31 USD
102515-556 102515-558
Human Recombinant BCMA (from HEK293 cells)
Proteins and Peptides
BCMA is a receptor for BAFF and APRIL. It promotes B cell survival and plays a role in the regulation of humoral immunity. Its downstream signaling is dependent on NF-kappaB and JNK.