Human Recombinant GITR (from HEK293 Cells)
Supplier: 0000027161
AG-40B-0028-C050 AG-40B-0028-3050
102515-466EA 443.08 USD
102515-466 102515-468
Human Recombinant GITR (from HEK293 Cells)
Proteins and Peptides
GITR (Glucocorticoid-induced TNF receptor) is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T cell receptor-mediated cell death. GITR serves as a negative regulator for NK cell activation. It mediates NF-kappaB activation via the TRAF2/NIK pathway.